Learn More
Enzo Life Sciences Seratrodast (10 mg). CAS: 112665-43-7
SDP
Supplier:  Enzo Life Sciences BMLRA1190010
Encompass_Preferred
Seratrodast is a potent and selective thromboxane A2 receptor (TP) antagonist with antioxidant properties. Purity: ≥98%. Solubility: Soluble in DMSO (12mg/ml); Ethanol (10mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1725
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.