Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103311 Supplier Invitrogen™ Supplier No. PA5103311
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SERF2.

SERF2 (small EDRK-rich factor 2) is the human ortholog of the C/ elegans MOAG-4 gene. Serf2 (small EDRK-rich factor 2) is the human ortholog of the C. elegans MOAG-4 gene. MOAG-4 was shown to be a positive regulator of protein aggregates. Inactivation of MOAG-4 leads to a rescue from proteotoxcity, and could play a role in human diseases such as Alzheimer's disease and Parkinson's disease. In addition, SERF2 can induce huntingtin aggregation in SK-N-SH neuroblastoma cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERF2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SERF2
Gene Accession No. P84101, P84102
Gene Alias 4F5REL; 4F5rel (4F5 related); C80083; FAM2C; Gastric cancer-related protein VRG107; H4F5REL; HsT17089; m4F5rel; Msmac1l; Protein 4F5-related; SERF2; Small EDRK-rich factor 2
Gene Symbols SERF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SERF2(Accession P84101), corresponding to amino acid residues M1-L31.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10169, 378702
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.