Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Serglycin Monoclonal Antibody (C1), Invitrogen™

Catalog No. PIMA543587
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543587 20 μL
PIMA543586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543587 Supplier Invitrogen™ Supplier No. MA543587
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This gene encodes a protein best known as a hematopoietic cell granule proteoglycan. Proteoglycans stored in the secretory granules of many hematopoietic cells also contain a protease-resistant peptide core, which may be important for neutralizing hydrolytic enzymes. This encoded protein was found to be associated with the macromolecular complex of granzymes and perforin, which may serve as a mediator of granule-mediated apoptosis. Two transcript variants, only one of them protein-coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serglycin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SRGN
Gene Accession No. P04917, P10124
Gene Alias gp600; hematopoetic proteoglycan core peptide; hematopoetic proteoglycan core protein; hematopoietic proteoglycan core protein; Mastocytoma proteoglycan core protein; p.PG; Platelet proteoglycan core protein; PPG; Prg; Prg1; proteoglycan 1, secretory granule; proteoglycan protein core for mast cell secretory granule; proteoglycan, secretory granule; secretory granule proteoglycan core peptide; secretory granule proteoglycan core protein; Serglycin; serglycin proteoglycan; Sgc; Srgn
Gene Symbols SRGN
Host Species Mouse
Immunogen Asp119-Gly148 (Accession P10124), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5552, 56782
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.