Learn More
Description
- Epredia™ Laser Cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Comes in multiple different colors with color-matched lids
- Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Comes in taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes with separate lids
- Not compatible with thermal tape style printers
Specifications
Specifications
| Pore Style | Small Squares |
| Quantity | 40 Cassettes/Taped Stack, 25 Stacks/Cs. |
| Closure Included | Yes |
| Closure Type | Separate Lid with Tab |
| Color | White |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.