Learn More
Kemtec™ Series and Parallel Circuits Kit
SCI_ED
Catalog No. S27538 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Includes:
D-Cell Batteries
No batteries
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27538 No batteries
S27537 D-Cell Batteries
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27538 Supplier Kemtec™ Supplier No. 1195
Add to Cart
Add to Cart
Kemtec™ Series and Parallel Circuits Kit teaches students to build parallel and series circuits using electrical components.
  • Students begin by following the flow of energy from a battery as it converts to chemical, electrical, light, sound, motion, and heat energy.
  • Students observe how energy flow differs when using batteries and loads in series and in parallel.
  • Allow for 5 hours of instruction and 5 hours of lab time.
  • Meets National Standards for Grades 3 to 6.
  • Kit includes:
    • Battery Holders
    • Motors
    • Light Bulbs
    • Light Bulb Holders
    • Alligator Clip Leads
    • Buzzers
    • Masking Tape
    • Propeller
    • Optional D-Cell Batteries
Specifications
Grade 3 to 6
Age 8 to 12
Includes No batteries
Weight (English) 5 lb.
Dimensions (L x W x H) 13 x 12 x 5 in.