Learn More
Cytiva Series S Sensor Chip Protein L 1-p
Supplier:  Cytiva 29205138
Encompass
A Biacore Extend sensor chip for site-directed antibody capture. Suitable for study of a wide range of antibody fragments including Fabs and dAbs.
Catalog No. 50-152-8207