Learn More
Cytiva SERIES S SENSOR CHIP CM3 CERTI
Supplier:  Cytiva BR100536
Encompass
An alternative to Sensor Chip CM5 with shorter dextran matrix and similar charge density - to explore alternative assay conditions.
Catalog No. 50-105-5322