Learn More
Cytiva SERIES S SENSOR CHIP CM4 1/PK
Catalog No. 501055512 Shop All Cytiva Products
Encompass
Use when sample contaminants have a high postive charge.
Catalog No. 50-105-5512 Supplier Cytiva Supplier No. 29104989
Use when sample contaminants have a high postive charge.