Learn More
Cytiva Series S Sensor Chip CM5, pack of 10
Catalog No. 501055509 Shop All Cytiva Products
Encompass
Series S Sensor Chip CM5, pack of 10, The most versatile chip available the first choice for immobilization via -NH2, -SH, -CHO, -OH or -COOH groups.
Catalog No. 50-105-5509 Supplier Cytiva Supplier No. 29149603
Series S Sensor Chip CM5, pack of 10, The most versatile chip available the first choice for immobilization via -NH2, -SH, -CHO, -OH or -COOH groups.