Learn More
Cytiva Series S Sensor Chip PEG
Catalog No. 501528217 Shop All Cytiva Products
Encompass
Biacore Series S Sensor Chip PEG enables interaction analysis that cannot otherwise be performed on dextran-based surfaces.
Catalog No. 50-152-8217 Supplier Cytiva Supplier No. 29239810
Biacore Series S Sensor Chip PEG enables interaction analysis that cannot otherwise be performed on dextran-based surfaces.