Learn More
Cytiva SERIES S SENSOR CHIP SA (1/PK)
Catalog No. 501055516 Shop All Cytiva Products
Encompass
For stable and convenient immobilization of biotinylated molecules. The most versatile chip available the first choice for immobilization via -NH2, -SH, -CHO, -OH or -COOH groups. Suitable for ligand fishing. Suitable for high capacity capture. Supports a wide range of immobilization levels. Attach proteins, nucleic acids, carbohydrates or small molecules.
Catalog No. 50-105-5516 Supplier Cytiva Supplier No. 29104992
For stable and convenient immobilization of biotinylated molecules. The most versatile chip available the first choice for immobilization via -NH2, -SH, -CHO, -OH or -COOH groups. Suitable for ligand fishing. Suitable for high capacity capture. Supports a wide range of immobilization levels. Attach proteins, nucleic acids, carbohydrates or small molecules.