Learn More
Cytiva Series S Sensor Chip SA, pack of 3
Catalog No. 501055318 Shop All Cytiva Products
Encompass
For stable and convenient immobilization of biotinylated molecules.
Catalog No. 50-105-5318 Supplier Cytiva Supplier No. BR100531
For stable and convenient immobilization of biotinylated molecules.