Learn More
Cytiva SERIES S SENSOR PROTEIN A 1PK
Supplier:  Cytiva 29127555
Encompass
Series S Sensor Chip Protein A, Pack of 1, Ensures orientation-specific binding of antibodies through the Fc portion.
Catalog No. 50-105-5503