Learn More
Johannes Lieder Series for Secondary Schools, Set No. IV. Hormone Organs and Hormonal Functions
SCI_ED
Catalog No. S02820 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Catalog No. Mounting Type
S02820 Cross and Longitudinal Section
1 options
Catalog No. S02820 Supplier Johannes Lieder Supplier No. 4470
Add to Cart
Add to Cart
4470 Series for Secondary Schools, Set No. IV. Hormone Organs and Hormonal Functions, 7 microscope slides
  • 4470 Set No. IV. Hormone Organs and Hormonal Functions.
  • 7 Microscope Slides
    1. Ovary of cat, with follicles and corpus luteum t.s.
    2. Testis of mouse, t.s. showing Leydig's cells
    3. Adrenal (suprarenal) gland of cat, t.s.
    4. Pancreas of cat, t.s. with islets of Langerhans,
    5. Thyroid gland, normal function t.s.
    6. Thyroid gland, over-activity of the gland t.s.
    7. Hypophysis (pituitary body) sagittal l.s.
Specifications
Classification Histology
Material Elastin Stained Glass
Mounting Type Cross and Longitudinal Section
Type Microscope Slides
Slide Type Prepared Slide