Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Seripettor™ Pro Pump Assembly

Catalog No. 03388506 Shop All BrandTech Scientific Products
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
03-388-506 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-388-506 Supplier BRANDTECH™ BRAND™ Supplier No. 704548
Add to Cart
Add to Cart

Accessories for Seripettor™ and seripettor™ pro Bottletop Dispensers.

  • Seripettor™ pump assembly provided with stainless steel lifting spring.
  • Seripettor™ pro pump assembly provided with Hastaloy™ (stainless) lifting spring, with PEI for UV protection.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Pump Assembly
Autoclavable Not Autoclavable
Color Dark Blue, Red, Translucent Amber
Volume (Metric) 10 mL
For Use With (Application) Dispensing Liquid
For Use With (Equipment) seripettor™ pro, 10 mL
Includes Pump assembly only
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.