Learn More
BRANDTECH™ seripettor™ pro Pump Assembly
Catalog No. 03388508 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Volume (Metric):
2 mL
Catalog No. Volume (Metric)
03-388-508 2 mL
1 options
Catalog No. 03-388-508 Supplier BRANDTECH™ Supplier No. 704551
Add to Cart
Add to Cart
For seripettor™ pro actuating unit
  • PPO
  • PEI (UV protection)
  • Hastaloy lifting spring
  • TRUSTED_SUSTAINABILITY
    Specifications
    Product Type Pump Assembly
    Volume (Metric) 2 mL