Learn More
Poltex Serological Pipette Holder with Door
SDP
Catalog No. 22000581
Change view
Click to view available options
Mounting:
Countertop Placement
Magnet Mount
Includes:
Holder Only
Pipette Controller Bracket
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Includes
22-000-581 Countertop Placement Holder Only
22-000-582 Magnet Mount Holder Only
22-000-583 Countertop Placement Pipette Controller Bracket
22-000-584 Magnet Mount Pipette Controller Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-000-581 Supplier Poltex Supplier No. DRSRPIP
Only null left
Add to Cart
Add to Cart
The Serological Pipette Holder with Door can be customized to meet your needs with different mounting options and hinge mounting for either side.
  • Door latches shut with magnetic catch.
  • Hinges can mount to either side.
  • Choose from powerful magnet mount (magnets mount to either side) or countertop placement.
  • Add a pipette filler bracket to any version (bracket mounts to either side).
  • See image for dimensions.
  • 1/4 in. thick PETG construction.
Specifications
Includes Holder Only
Material PETG
Mounting Countertop Placement