Learn More
Eurofins Discoverx Serotonin 5-HT, 10 mg. (92-1108)
Supplier:  Eurofins Discoverx 921108
Encompass
Serotonin, or 5-hydroxytryptamine (5-HT), is a mono-amine neurotransmitter derived from tryptophan that is responsible for regulation of mood and associated with depression. An agonist for the serotonin (HRT) receptors, it is primarily located in the CNS and GI tract, and modulates physiological functions related to neurotransmission, GI motility, pain perception, hemostasis, hunger, and cardiovascular integrity. Provided as 10 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9643
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.