Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A9/Centerin Antibody [DyLight 755], R&D Systems™
SDP

Catalog No. AF5429Z
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5429Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF5429Z Supplier R&D Systems Supplier No. AF5429Z

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

Serpin A9/Centerin Polyclonal specifically detects Serpin A9/Centerin in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Serpin A9/Centerin
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias GCET1, SERPINA11, SERPINA11b
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A9, Ala24-Thr417, Accession # AAQ89063
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 327657
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.