Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Serpin E1/PAI-1 Antibody Pair [HRP]
SDP

Catalog No. NB010159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010159 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010159 Supplier Novus Biologicals™ Supplier No. NBP27956015PLATES
Only null left
Add to Cart
Add to Cart

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Serpin E1/PAI-1.

Specifications

Assay Range 46.88 pg/mL
Assay Sensitivity 46.88-3000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Serpin E1/PAI-1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Gene ID (Entrez) 5054
Gene Symbol SERPINE1
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 125 ng of recombinant Human Serpin E1/PAI-1.
  • Capture Antibody: 1 mg/mL of mouse anti-Human Serpin E1/PAI-1 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Serpin E1/PAI-1 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target Serpin E1/PAI-1
    Technique 1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.03125 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.