Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin E1/PAI-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067868
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1786SP 25 μg
MAB1786100 100 μg
MAB1786500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1786SP Supplier R&D Systems Supplier No. MAB1786SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Serpin E1/PAI-1
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 242816
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 2.5 ug/mL, Neutralization 0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P05121
Gene Alias Endothelial plasminogen activator inhibitor, Nexin, PAI1, PAI-1, PAI1PAI-1, PAISerpin E1, PLANH1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Gene Symbols SERPINE1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin E1/PAI-1 Met1-Pro402 Accession # P05121
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5054
Test Specificity Detects human Serpin E1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Serpin A1, A3, A4, A8, C1, F1, F2, I1, I2, recombinant mouse Serpin D1 or E2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.