Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA3G Monoclonal Antibody (MoFo 29.2)
GREENER_CHOICE

Catalog No. AMP030 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMP-030 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMP-030 Supplier Invitrogen™ Supplier No. MA1030
  • $620.58
    $481.65 / Each of 1

    Save $138.93

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Armenian Hamster Monoclonal Antibody

MA1-030 detects Spi2A in human, mouse and hamster samples. MA1-030 has been used successfully in Western blot, ICC/IF and ELISA procedures. By Western blot this antibody detects a ~49 kDa band representing the Spi2A protein. The MA1-030 immunogen is a synthetic peptide corresponding to residues N(418) P E R S M N F P N G E G A S S Q R L C(436) of hamster Spi2A protein.

Serine protease inhibitor 2A, or Serpin 2A (spi2A), inhibits lysosomal executioner proteases, thus enhancing the survival of cytotoxic T lymphocytes and allowing them to develop into memory CD8 T cells. Through its function of preserving T cells, spi2A has become a research target for such topics as immunity, tumor cell, and AIDS research.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA3G
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MoFo 29.2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Serpina3g
Gene Accession No. Q5I2A0
Gene Alias 2A2; AI119734; alpha-1 antiproteinase; serine (or cysteine) peptidase inhibitor, clade A, member 3G; serine (or cysteine) proteinase inhibitor, clade A, member 3G; serine protease inhibitor 2-1; serine protease inhibitor 2A; Serine protease inhibitor A3G; serpin 2A; Serpin A3G; Serpina3g; spi2; Spi2/eb.1; Spi2-1; Spi2A
Gene Symbols Serpina3g
Host Species Armenian Hamster
Immunogen Synthetic peptide corresponding to residues N(418) P E R S M N F P N G E G A S S Q R L C(436) of hamster Spi2A protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20715
Target Species Human, Mouse, Hamster
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.