Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571456 Supplier Invitrogen™ Supplier No. PA571456
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 91%; Horse: 91%; Human: 91%; Mouse: 91%; Pig: 91%; Rat: 91%.

SERPINH1 is a member of the serpin superfamily of serine proteinase inhibitors, and its expression is induced by heat shock. The SERPINH1 protein localizes to the endoplasmic reticulum lumen and binds collagen; thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to SERPINH1 have been found in patients with rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINH1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SERPINH1
Gene Accession No. P19324, P50454
Gene Alias 47 kDa heat shock protein; arsenic-transactivated protein 3; AsTP 3; AsTP3; BERF-1; beta enolase repressor factor 1; CBP 1; CBP 2; CBP1; CBP2; Cell proliferation-inducing gene 14 protein; collagen binding protein 1; collagen binding protein 2; collagen-binding protein; Colligen; Colligin; Colligin 1; Colligin 2; colligin-1; colligin-2; GP46; heat shock protein 47; HGNC 1547; Hsp 47; HSP47; J6; OI10; PIG 14; PIG14; PPROM; RA A47; RA-A47; rheumatoid arthritis antigen A-47; rheumatoid arthritis-related antigen RA-A47; serine (or cysteine) peptidase inhibitor, clade H, member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2); serine (or cysteine) proteinase inhibitor, clade H, member 1; serine protease inhibitor J6; serine proteinase inhibitor, clade H (heat shock protein 47), member 1; serpin family H member 1; serpin H1; serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serpin peptidase inhibitor, clade H, member 1; SERPINH1; SERPINH2
Gene Symbols SERPINH1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human SERPINH1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12406, 871
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.