Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINI2 Recombinant Rabbit Monoclonal Antibody (018), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
$310.00 - $593.00
Specifications
| Antigen | SERPINI2 |
|---|---|
| Clone | 018 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA543425
|
Invitrogen™
MA543425 |
50 μL |
Each for $310.00
|
|
|||||
PIMA543141
![]() |
Invitrogen™
MA543141 |
100 μL |
Each for $593.00
|
|
|||||
Description
Specificity: Human SerpinI2
The protein encoded by this gene is a member of the serine protease inhibitor superfamily made up of proteins which play central roles in the regulation of a wide variety of physiological processes, including coagulation, fibrinolysis, development, malignancy and inflammation. The gene product may have a role in a growth-control, possibly growth-suppressing pathway and, when impaired, may be involved in pancreatic carcinogenesis. The protein is a member of the plasminogen activator inhibitor-1 family, a subset of the serpin superfamily whose members act as tissue-specific tPA inhibitors. Two alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.Specifications
| SERPINI2 | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| MEPI; Myoepithelium-derived serine protease inhibitor; neuroserpin; PANCPIN; Pancreas-specific protein TSA2004; pancreas-specific serpin; peptidase inhibitor 14; PI14; PI-14; protease inhibitor 14; serine (or cysteine) peptidase inhibitor, clade I, member 2; serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 2; serine (or cysteine) proteinase inhibitor, clade I (pancpin), member 2; serine protease inhibitor 14; serpin family I member 2; serpin I2; serpin peptidase inhibitor, clade I (pancpin), member 2; serpin peptidase inhibitor, clade I, member 2; SERPIN12; SERPINI2; Spi14; TSA2004 | |
| SERPINI2 | |
| IgG | |
| Protein A | |
| Antibody |
| 018 | |
| ELISA, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O75830 | |
| 5276 | |
| Recombinant Human SerpinI2 protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| SERPINI2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title