Learn More
Med Vet International Simparica Trio Chewable Tablets For Dogs 44.1 To 88 Pounds, Green Label, 6 Dose/Box
Supplier:  Med Vet International RXSIMPARICATRIO1GR
Encompass_Preferred
Simparica TRIO (sarolaner, moxidectin, and pyrantel) Chewable Tablets prevent heartworm disease caused by Dirofilaria immitis, kills adult fleas (Ctenocephalides felis) and is indicated for the treatment and prevention of flea infestations, the treatment and control of tick infestations with Amblyomma americanum (lone star tick), Amblyomma maculatum (Gulf Coast tick), Dermacentor variabilis (American dog tick), Ixodes scapularis (black-legged tick), and Rhipicephalus sanguineus (brown dog tick), and the treatment and control of roundworm (immature adult and adult Toxocara canis and adult Toxascaris leonina) and adult hookworm (Ancylostoma caninum and Uncinaria stenocephala) infections for one month in dogs and puppies 8 weeks of age and older, and weighing 2.8 pounds or greater. Simparica TRIO is indicated for the prevention of Borrelia burgdorferi infections as a direct result of killing Ixodes scapularis vector ticks. See package insert for complete product details. EXPIRATION 4/27
Catalog No. 50-282-8575
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.