Learn More
Serum Amyloid A1/A2 Antibody (SAA/2868R), DyLight 594, R&D Systems™

Description
Specifications
Specifications
| Antigen | Serum Amyloid A1/A2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SAA/2868R |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | PIG4;SAA;SAA1;SAA2;Serum amyloid A-1 protein;Serum amyloid A-2 protein;TP53I4 |
| Host Species | Rabbit |
| Immunogen | Recombinant full-length human Serum Amyloid A1/A2 (SAA) protein |
| Purification Method | Protein A or G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.