Learn More
STEMCELL Technologies EasySep™ Mouse Pan-DC Enrichment Kit II, For processing: 2 x 10^9 cells
Supplier: STEMCELL Technologies 19863
The EasySep™ Mouse Pan-DC Enrichment Kit II is designed to isolate dendritic cells (DCs) from mouse splenocytes by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.This product replaces the EasySep™ Mouse Pan-DC Enrichment Kit (Catalog #19763), for improved yield and even faster cell isolations.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.