Learn More
Beckman Coulter Beckman Coulter Serum Protein Multi-Calibrator, 6 x 2 mL (six levels) - for AU-series and compatible analyzers
Supplier:  Beckman Coulter OSR3021
Encompass
Six-level serum protein multi-calibrator supplied as six 2 mL levels for calibration of immuno-turbidimetric serum protein assays on compatible clinical chemistry analyzers.
  • Prepared from human serum with constituent values adjusted.
  • Provides six calibration levels across clinically relevant concentrations.
  • Covers common serum protein analytes including IgG, IgA, IgM, complement C3 and C4, transferrin, C-reactive protein, anti-streptolysin O, and ferritin.
  • Packaged as six 2 mL levels for multi-point calibration workflows.
  • Designed for use with immuno-turbidimetric reagents on AU-series and compatible analyzers.
Catalog No. NC3453468