Learn More
METTLER TOLEDO™ Service for Advanced MS and ML Balances
Professional installation completed by a trained service technician will guarantee maximum performance and optimal use.
$489.48 - $1721.53
Specifications
Duration 1 Day
Product Type Balance Service
View More Specs
Products 8
Catalog Number Mfr. No. Includes For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Includes For Use With (Equipment) Price Quantity & Availability  
01-670-007
View Documents
METTLER TOLEDO™
B99484130102
IPac Balances with Accredited ACC Advanced MS, ML Precision Balances
Each for $1,146.40
Only null left
Add to Cart
01-670-016
View Documents
METTLER TOLEDO™
B99484130101
IPac Balances with Accredited ACC Advanced MS, ML Analytical Balances
Each for $1,146.40
Only null left
Add to Cart
01-670-017
View Documents
METTLER TOLEDO™
B99504130102
EQPac Balance with Accredited ACC Advanced MS, ML Precision Balances
Each for $1,672.27
Only null left
Add to Cart
01-670-023
View Documents
METTLER TOLEDO™
B99494130101
EQPac Regulated Balances with USP Advanced MS, ML Analytical Balances
Each for $1,721.53
Only null left
Add to Cart
01-670-029
View Documents
METTLER TOLEDO™
B99504130101
EQPac Balance with Accredited ACC Advanced MS, ML Analytical Balances
Each for $1,672.27
Only null left
Add to Cart
01-670-031
View Documents
METTLER TOLEDO™
B99494130102
EQPac Regulated Balances with USP Advanced MS, ML Precision Balances
Each for $1,721.53
Only null left
Add to Cart
01-670-033
View Documents
METTLER TOLEDO™
B99474130101
StarterPac Installation with ACC Advanced MS, ML Analytical Balances
Each for $489.48
Only null left
Add to Cart
01-670-037
View Documents
METTLER TOLEDO™
B99474130102
StarterPac Installation with ACC Advanced MS, ML Precision Balances
Each for $489.48
Only null left
Add to Cart
Description
Description
To achieve peak performance and compliance with quality systems and regulatory requirements, take the right steps at the beginning of equipment use. The Accuracy Calibration Certificate (ACC) is the new benchmark in the calibration of weighing equipment. Tests include repeatability, eccentricity and error of indication. A statement of measurement uncertainty is also included.
  • StarterPac includes: Professional installation performed by a trained and authorized service technician, installation confirmation/documentation, immediate deployment and use of your equipment, professionally trained operating personnel
  • IPac includes: Documented standard qualification and initial calibration, compliance with internal and external regulations, immediate deployment for quick, safe release to routine operation, and improved productivity with correct equipment set-up and initial user training
  • EQPac includes: Regulatory conformance qualification documentation, professional qualification including test procedures, specification of weighing ranges and calibration, immediate deployment for quick, safe release to begin routine operation, improved productivity with correct equipment set-up and initial user training, cost savings through the use of a manufacturer developed qualification protocol
Specifications
Specifications
1 Day
Balance Service
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.