Learn More
Mettler Toledo™ Service for Advanced MX Balances
Catalog No. 01258284 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Service Offering:
EQPac Balance with Accredited ACC
EQPac Regulated Balances with USP
IPac Balances with Accredited ACC
StarterPac Installation with ACC
For Use With (Equipment):
Advanced MX Analytical Balances
Advanced MX Precision Balances
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Service Offering For Use With (Equipment)
01-258-284 IPac Balances with Accredited ACC Advanced MX Analytical Balances
01-258-273 EQPac Balance with Accredited ACC Advanced MX Precision Balances
01-258-274 EQPac Balance with Accredited ACC Advanced MX Analytical Balances
01-258-278 EQPac Regulated Balances with USP Advanced MX Precision Balances
01-258-279 EQPac Regulated Balances with USP Advanced MX Analytical Balances
01-258-288 StarterPac Installation with ACC Advanced MX Precision Balances
01-258-283 IPac Balances with Accredited ACC Advanced MX Precision Balances
01-258-289 StarterPac Installation with ACC Advanced MX Analytical Balances
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-258-284 Supplier Mettler Toledo™ Supplier No. B99484130127
Add to Cart
Add to Cart
Professional installation completed by a trained service technician will guarantee maximum performance and optimal use.
To achieve peak performance and compliance with quality systems and regulatory requirements, take the right steps at the beginning of equipment use.
  • The Accuracy Calibration Certificate (ACC) is the new benchmark in the calibration of weighing equipment.
  • Tests include repeatability, eccentricity and error of idication.
  • A statement of measurement uncertainty is also included.
  • StarterPac includes: Professional installation performed by a trained and authorized service technician, installation confirmation/documentation, immediate deployment and use of your equipment, professionally trained operating personnel
  • IPac includes: Documented standard qualification and initial calibration, compliance with internal and external regulations, immediate deployment for quick, safe release to routine operation, and improved productivity with correct equipment set-up and initial user training
  • EQPac includes: Regulatory conformance qualification documentation, professional qualification including test procedures, specification of weighing ranges and calibration, immediate deployment for quick, safe release to begin routine operation, improved productivity with correct equipment set-up and initial user training, cost savings through the use of a manufacturer developed qualification protocol
  • Extended Warranty: Extends manufacturer's warranty by an additional 12 months
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Advanced MX Analytical Balances
Product Type Balance Service
Service Offering IPac Balances with Accredited ACC