Learn More
Labconco™ Service Fixture Kits
SDP
Catalog No. 09039049 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Cold Water (CW) GN Fixture Kit
Cold Water (CW) Service Fixture Kit
Gas (GAS) Service Fixture Kit
HW/CW Mixing GN Fixture Kit
Nitrogen (NIT) Service Fixture Kit
Oxygen (OXY) Service Fixture Kit
Steam (STM) Service Fixture Kit
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
09-039-049 Cold Water (CW) GN Fixture Kit
09-039-046 Steam (STM) Service Fixture Kit
09-039-047 Nitrogen (NIT) Service Fixture Kit
09-039-048 Oxygen (OXY) Service Fixture Kit
09-039-050 HW/CW Mixing GN Fixture Kit
09-039-039 Cold Water (CW) Service Fixture Kit
09-039-042 Gas (GAS) Service Fixture Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 09-039-049 Supplier Labconco™ Supplier No. 9827900
Add to Cart
Add to Cart
Kit includes copper tubing, brass valve, knob handle and outlet.
  • Max. working pressure of 40 psi.
  • Kits mount on the right or left side of any Protector Premier, XStream, XL, or PVC Hood.
  • Each kit includes one remotely-controlled service fixture with valve and tubing, color-coded fixture knob.
Specifications
For Use With (Equipment) Protector XLE Laboratory Hood
Weight (English) 10 lb.
Weight (Metric) 4.5 Kg
Includes 0.37 in. dia. Brass Valve, Copper Tubing, Green Fixture Knob, Hose Connector
Product Type Cold Water (CW) GN Fixture Kit
For Use With (Application) Cold water service
WARNING: Reproductive Harm - www.P65Warnings.ca.gov