Learn More
Honeywell™ Servus™ Neoprene III with Neo-Grip Soles
Description
- Unique dipping process makes boots soft and pliable for less leg fatigue
- Neo-Grip™ outsoles provide excellent traction on wet surfaces to reduce injuries from slips and falls
- Removable Breathe-O-Prene™ insoles provide shock absorption and moisture-wicking comfort
- Tough, comfortable fabric lining
- Available in copper tan Mid Pac and Hi Pac
- Steel toe
Mid Pac
- Height is 30.48cm (12 in.)
- US men's sizes 6-13
- Height is 40.6cm (16 in. )
- US men's sizes 6, 8-9, 11, 13
Compliance
Meets electrical hazard requirements of ASTM F2413-05. Steel toe also meets ASTM F2413-05. Neoprene styles meet ASTM F2413-11 EH
Specifications
Specifications
| Certifications/Compliance | ANSI Z41 PT 99 |
| Color | Tan |
| Height (English) | 12 in. |
| Height (Metric) | 30.48 cm |
| Size | 13 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.