Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Servus™ Neoprene III with Neo-Grip Soles

Catalog No. 19038409
Change view
Click to view available options
Size:
6
7
8
9
11
13
Height (Metric):
30.48 cm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (Metric)
19-038-409 9 30.48 cm
19-081244 6 30.48 cm
19-019518 8 30.48 cm
19-019-535 13 30.48 cm
19-044089 7 30.48 cm
19-044079 11 30.48 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-038-409 Supplier Honeywell™ Supplier No. 22148CTM090

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

100% waterproof seamless construction provides comfort, flexibility and excellent traction

  • Unique dipping process makes boots soft and pliable for less leg fatigue
  • Neo-Grip™ outsoles provide excellent traction on wet surfaces to reduce injuries from slips and falls
  • Removable Breathe-O-Prene™ insoles provide shock absorption and moisture-wicking comfort
  • Tough, comfortable fabric lining
  • Available in copper tan Mid Pac and Hi Pac
  • Steel toe

Mid Pac

  • Height is 30.48cm (12 in.)
  • US men's sizes 6-13
  • Hi Pac
    • Height is 40.6cm (16 in. )
    • US men's sizes 6, 8-9, 11, 13

    Compliance

    Meets electrical hazard requirements of ASTM F2413-05. Steel toe also meets ASTM F2413-05. Neoprene styles meet ASTM F2413-11 EH

    TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI Z41 PT 99
    Color Tan
    Height (English) 12 in.
    Height (Metric) 30.48 cm
    Size 9
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.