Learn More
Honeywell™ Servus™ PRM Safety Knee Boot
Description
- Flexible steel midsoles provide under-foot puncture protection
- Open pattern outsole design self-cleans liquid and debris with every step, helping to prevent slip and trip hazards
- Removable FOOT FORM contour cushion insoles provide all-day comfort and support
- Available in US men sizes 7 to 14
- Hi Boot (38.1cm/15 in.)
- Steel toe
Compliance
Meets ASTM F2413-11 M I/75 C/75 and PR.
Specifications
Specifications
| Height (English) | 15 in. |
| Size | 13 |