Learn More
Honeywell™ Servus™ Puncture-Resistant Midsole
Catalog No. 199201763
Change view
Click to view available options
Size:
8
9
10
11
12
13
14
15
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-920-1763 10
19-920-1761 8
19-920-1762 9
19-920-1764 11
19-920-1765 12
19-920-1767 14
19-920-1768 15
19-920-1766 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-920-1763 Supplier Honeywell™ Supplier No. 29000BLK100
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Cushion-covered, flexible steel inserts provide puncture protection underfoot
  • Use as an insole or midsole
  • Will fit most over-the-foot boots
TRUSTED_SUSTAINABILITY
Specifications
Material Cushion
Size 10