Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Servus™ Ranger™ Flexible Metatarsal Guard Boots

Catalog No. 19024927
Change view
Click to view available options
Size:
6
7
8
9
12
13
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-024927 7
19-024926 6
19-024931 12
19-024928 8
19-024929 9
19-044698 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-024927 Supplier Honeywell™ Supplier No. 2149ODM070
This item is not returnable. View return policy

Rugged Flex-Gard construction provides longer-life footwear without sacrificing comfort

Specialized compound prevents ozone deterioration for longer life footwear.

  • Ultra-composite shank is longer, wider and positioned for optimal support to help prevent foot pain
  • SafeStep™ puncture-resistant steel midsoles provide maximum puncture protection
  • Safe-Toe™ steel toe protects worker from crush and impact hazards
  • Removable Foot Form™ contour cushion insoles support and wick away moisture
  • Heavy Trac Tread sole, heel and knurled protective overlays are tough and durable

  • Available in Mid Pac, men's sizes 6-10, 12-13
  • Height: 30.5cm (12 in.)
  • Closure: Buckle
  • Color: Olive

Compliance

Meet ASTM F2413-11 M I/75 C/75 Mt/75 and PR

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Size 7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.