Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Servus™ SF™ Chem Overboots

Catalog No. 19122573
Change view
Click to view available options
Size:
Small, 6 to 8
X-Large, 14 to 15
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-122-573 Small, 6 to 8
19-122-573C X-Large, 14 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-122-573 Supplier Honeywell™ Supplier No. 11095BLMSML
This item is not returnable. View return policy

Fit over all types of shoes and boots

Provide general chemical resistance for a variety of applications, including WMD and incident response.

  • Super-fit construction
  • Slip-coated interiors
  • Slip-resistant outsoles
  • Seamless, one-piece construction with molded buttons
  • No swelling or softness from exposure to petroleum distillates
  • Height: 12 in. (30.5cm)
  • Made in USA

Compliance

Tested to the chemical permeation requirements of MIL-O-43995C and CID A-A-59520.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested to the chemical permeation requirements of MIL-O-43995C and CID A-A-59520.
Color Black
For Use With (Application) Provide general chemical resistance for a variety of applications, including WMD and incident response.
Height (English) 12 in.
Height (Metric) 30.5 cm
Seam Type Seamless
Size Small, 6 to 8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.