Learn More
Sesame Oil, NF, Spectrum™ Chemical
SDP
Catalog No. 18606189 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
6 x 500 mL
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-189 4 x 4 L Amber Glass Bottle
18-606-187 6 x 500 mL Amber Glass Bottle
18-606-191 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-189 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SE1304LTGLCS4
Add to Cart
Add to Cart
SE130, 8008-74-0
Spectrum™ Chemical Sesame Oil, NF is used as an excipient solvent carrier in suspensions and emulsions, ointments, liniments, ophthalmic preparations, oral capsules and suppositories. All Spectrum Chemical NF are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
CAS 8008-74-0
Specifications
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 4 x 4 L
Saponification Value 188-195