Learn More
3M™ Sesame Protein ELISA Kit, E96SES, Sold by AquaPhoenix Scientific
Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
96-well plate
Description
- Designed to detect processed and unprocessed target allergen proteins
- Test can accommodate environmental swab, CIP rinse water and food samples
- Universal extraction protocol streamlines testing for multiple analytes, saving reagents and time
Specifications
Specifications
Detectable Analytes | Sesame protein |
Product Type | ELISA kit |
Quantity | 96-well plate |
Test Time | 50 min. |