Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SESN3 Native Protein
SDP

Catalog No. NBP144988 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144988 0.15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144988 Supplier R&D Systems™ Supplier No. NBP144988

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

Specifications

For Use With (Application) Western Blot
Formulation SDS PAGE sample buffer
Gene ID (Entrez) 143686
Quantity 0.15 mL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias SEST3MGC29667, sestrin 3, sestrin-3
Gene Symbol SESN3
Inhibitor Properties Western blotting positive control for SESN3
Research Category Cell Cycle and Replication
Product Line Novus
Purity or Quality Grade >85%, by HPLC
Protein SESN3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.