Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SESN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522220 Supplier Invitrogen™ Supplier No. PA522220
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, Raw264.7, C2C12, Rat2. Predicted reactivity: Mouse (94%), Rat (93%), Xenopus laevis (80%), Rhesus Monkey (98%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May function as an intracellular leucine sensor that negatively regulates the TORC1 signaling pathway (PubMed:25263562). May also regulate the insulin-receptor signaling pathway through activation of TORC2. This metabolic regulator may also play a role in protection against oxidative and genotoxic stresses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SESN3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SESN3
Gene Accession No. P58005, Q9CYP7
Gene Alias 5630400E15Rik; BC003348; Sesn3; SEST3; sestrin 3; sestrin-3
Gene Symbols SESN3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SESN3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143686, 315427, 75747
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.