Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Set of 4 dNTPs (100mm)

Catalog No. MP1NTACG100 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
4 x 100 μmol
4 x 25 μmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1NTACG100 4 x 25 μmol
MP1NTACG111 4 x 100 μmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP1NTACG100 Supplier MP Biomedicals™ Supplier No. 11NTACG100
Add to Cart
Add to Cart

Offers maximum flexibility in preparation of reaction mixes. MP Biomedicals Set of 4 dNTPs (100mm) includes four separate vials of highly purified sodium salts of dATP, dCTP, dGTP and dTTP. These solutions achieve sensitive and consistent PCR results.

This product allows the addition of each dNTP to be altered as required for specific experiments, such as:

  • When adding a radioactive nucleotide for labeling by PCR
  • To increase the chance of non-specific mutations
  • In PCR, real-time PCR, cDNA synthesis and DNA sequencing
TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 mM
Content And Storage Set 4 DNTPs composed of four separate vials of sodium salts of dATP; dCTP; dGTP and dTTP. Store at -20°C.
Purity or Quality Grade 99+%
Quantity 4 x 25 μmol
For Use With (Application) Standard PCR, Real Time PCR (qPCR), RT-PCR
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.