Learn More
Description
- Convenient assortment of seven commonly used laboratory tools.
- Contains one each of the following items: 6-hole test tube rack, test tube brush, test tube clamp, spatula, crucible tongs, lab scoop, and forceps.
Specifications
Specifications
| Color Code | Gray |
| Color-coded | No |
| For Use With (Equipment) | Test Tubes, Crucibles, Bottles, and Beakers |
| Handle Material | Metal |
| Material | Stainless Steel, Wood, Nylon |
| Includes | 6-Hole Test Tube Rack (1), Test Tube Brush (1), Test Tube Clamp (1), Spatula (1), Crucible Tongs (1), Lab Scoop (1), and Forceps (1) |
| No. per Pack | 1 Set |
| Product Type | Lab Tools Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

