Learn More
Air-Tite Products SET™Inject Luer Slip Syringe
Catalog No. 14817263 Shop All Air-Tite Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-817-263 1 mL
14-817-265 2 mL
14-817-267 5 mL
14-817-262 10 mL
14-817-264 20 mL
14-817-266 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
NO
Catalog No. 14-817-263 Supplier Air-Tite Products Supplier No. RS1SET
Only null left
Add to Cart
Add to Cart
NO
SET™ inject 3-Part Syringes, manufactured in Turkey and 510K registered for human medical use, are designed for precision, safety, and comfort.
  • Made from polypropylene with glass-like transparency, they feature a latex-free isoprene rubber gasket and silicone oil lubrication for smooth piston movement, along with a stop-point mechanism to prevent accidental removal.
  • Their ergonomic design allows single-handed use, while high impermeability ensures accurate dosing.
  • Each syringe is sterilized with ethylene oxide and individually packed in medical-grade blister packs with transparent film.
Specifications
Autoclavable Not Autoclavable
Material Polypropylene (Barrel), Polypropylene (Plunger), Polyisoprene Rubber (Tip)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Syringe
Disposable Single-use
Volume (Metric) 1 mL
Connection Type Centric Luer Lock
Graduated Yes
Sterility Sterile
Not Registered with Health Canada for Human Medical Use