Learn More
Eisco™ Premium Lab Metalware Set
Catalog No. 501042880 Shop All Eisco Products
Change view
Click to view available options
Material:
metal
Catalog No. Material
501042880 metal
1 options
This item is not returnable. View return policy
Catalog No. 501042880 Supplier Eisco™ Supplier No. MTST1/RSD
This item is not returnable. View return policy
Includes
Base, rod, clamp, retort ring
Single package containing support equipment for any experiment.
  • Made from durable metal
  • Stainless-steel rod
  • Powder-coated support stand base
  • Four-pronged, cork-lined clamp and metal retort ring
Specifications
Material metal
Includes Powder-coated stand base, stainless-steel rod, four-pronged cork-lined clamp, retort ring
Product Type Lab Metalware Set
Description Contains support equipment for any experiment