Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Seven-Piece Tool Kit
SDPSCI_ED

Catalog No. S47246 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color Code:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
S47246 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S47246 Supplier United Scientific™ Supplier No. TKIT04
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Adjustable crescent wrench, 152 mm
  • Slip-joint pliers, 152 mm
  • Long-nose side-cutter pliers, 140 mm
  • Unbreakable plastic screwdriver handle with four blades (mechanic, Phillips and electrician types)

This Seven-Piece Tool Kit is high quality and functional.

  • Drop-forged tool kit is ideal for classroom projects.
  • Comes in a recloseable polybag.

Specifications

Closure Size Various
Color Code Orange
Color-coded No
For Use With (Equipment) Assembling or dismantling equipment
Handle Material Plastic
Length (English) Various
Length (Metric) Various
Length (Metric) Blade Various
Material Metal
For Use With (Application) Classroom projects
Includes 6 in. Slip joint pliers, 6.5 in. L nose pliers, 6 in. adjustable wrench, 3 piece offset screwdriver set, 6 in. screwdriver set featuring slotted bits, Phillips™ bits, and nut drivers
Handle Type Various
Product Type Tool Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.