Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Sextant, Brass
SDPSCI_ED

Catalog No. S00660 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00660 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00660 Supplier United Scientific™ Supplier No. SXTNT1
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Brass Sextant is the original GPS.

  • Designed for beginners as well as experienced navigators.
  • This sextant features an adjustable index mirror, index, and horizon shades.
  • Durable brass construction, and features like more expensive models make this a great value.
  • Weighs only 0.75 lb.
  • Includes activity guide.

Specifications

Grade 6 to 12
Format Kit
Material Brass
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.