Learn More
Cliawaived Inc Sexual health vaginal collection kit
SDP
Supplier:  Cliawaived Inc VSBPS00071511
Encompass
Sexual health vaginal collection kit for the first true PCR point-of-care device for STI that’s all-in-one and requires no separate instrument or reader. Results available during the patient’s visit help to eliminate the guesswork of empirical medicine and patient callbacks associated with lab send outs. With the confidence of a PCR result, clinicians can prescribe the correct treatment and engage in a clear, guiding discussion with the patient.Our sexual health test targets three pathogens that (Chlamydia/Gonorrhea/Trichomoniasis ), if left untreated, can cause permanent damage to a woman’s reproductive system, including infertility, or potentially causing fatal ectopic pregnancy (pregnancy that occurs outside the womb). Fast PCR can help with early detection of these serious infections.
Catalog No. NC2578601
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.