Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEZ6/BSRP-C Antibody (Abbv-011) [PE] - (Research Grade Biosimilar), R&D Systems™
SDP

Catalog No. NBP328127PE
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP328127PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP328127PE Supplier R&D Systems Supplier No. NBP328127PE

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Human Monoclonal Antibody

SEZ6/BSRP-C Monoclonal antibody specifically detects SEZ6/BSRP-C in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay

Specifications

Antigen SEZ6/BSRP-C
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Abbv-011
Conjugate PE
Formulation PBS
Gene Alias hSEZ-6, seizure related 6 homolog (mouse)
Host Species Human
Immunogen SEZ6
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124925
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.