Learn More
Lonza Walkersville SF Cell Line 4D-Nucleofector X Kit S
Supplier:  Lonza Walkersville V4XC2032
Encompass
The SF Cell Line 4D-Nucleofector X Kit S is one of our three 4D-Nucleofector Kits that is suited for transfecting difficult-to-transfect cell lines in the 20 µL 16-well Nucleocuvette Strip format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCells maintain functionality post transfectionConditions are transferable between 4D-Nucleofector System, 96-well Shuttle System and 384-well Nucleofector SystemTransferability to 4D-Nucleofector LV UnitApplications:Transfection of low cell numbers in the range of 5 x 10⁴ to 1 x 10⁶ cellsMedium throughput applicationsOptimization of transfection conditions e.g. substrate titrationsContent:1 x 0.675 mL SF Cell Line Nucleofector; Solution1 x 0.15 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)2 x 16-well Nucleocuvette Strips (20 μL)
Catalog No. NC0281112